penta-Sodium triphosphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-29-4
:
Formula:Na₅O₁₀P₃
MDL Number:MFCD00003514
:367.86 g/mol
Merck Index:14,8697
Melting Pt:622 °C
Density:2.52 g/cm³ (20 °C)
EINECS:231-838-7
:GHS07
Storage Temperature:Ambient