Cesium nitrate Cabot, high purity, Sigma-Aldrich®
Catalog # SIAL562521-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: Nitric acid cesium salt
:
:194.91 g/mol
Merck Index:12,02057
Boiling Pt:737 °C (1013 hPa)
Melting Pt:414 °C
Density:3.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-146-8
UN:1451
ADR:5.1,III
:M-Clarity™ quality level = MQ100
:GHS03
:GHS07
CAS Number:7789-18-6
Formula:CsNO₃
MDL Number:MFCD00010963