Cesium nitrate ≥99.999% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Nitric acid cesium salt
CAS Number:7789-18-6
:
Formula:CsNO₃
MDL Number:MFCD00010963
:194.91 g/mol
Merck Index:12,02057
Boiling Pt:737 °C (1013 hPa)
Melting Pt:414 °C
Density:3.68 g/cm³ (20 °C)
UN:1451
ADR:5.1,III
:M-Clarity™ quality level = MQ100
EINECS:232-146-8
:GHS03
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: