Potassium iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:686 °C
Density:3.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-659-4