Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium iodide

Potassium iodide

Supplier:   Lovibond
Potassium iodide
Supplier:  Lovibond
Certificates
Potassium iodide

2 Options Available Below

Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:686 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 250
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 27,70 €
    27,70 €
    1 * 250 ST
       27,70 €

  • 100 Tab.
    Log in for availability
    Log in for availability
    1 * 100 Tablet
    1 * 100 Tablet - 20,70 €
    20,70 €
    1 * 100 Tablet
       20,70 €