Order Entry
Germany
ContactUsLinkComponent
Potassium hexacyanoferrate(III), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Environment
Potassium hexacyanoferrate(III), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Environment
Potassium hexacyanoferrate(III), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    464 g/l (20 °C)
  • Molar Mass:
    329.25 g/mol
  • Bulk Density:
    900 - 1000 kg/m3
  • Density:
    1.85 g/cm3 (20 °C)
  • pH:
    6 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together