Potassium carbonate, Sigma-Aldrich®
Catalog # SIALP1472-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
CAS Number:584-08-7
Formula:K₂CO₃
MDL Number:MFCD00011382