Potassium carbonate ≥98%, powder -325 mesh, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:13,07702
Boiling Pt: Dec.
Melting Pt:891 °C
Density:2.430 g/cm³ (19 °C)
EINECS:209-529-3
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Form:powder
- Assay:98.0 - 102.0 %
- Particle size distribution 50 %:Max. 44 µm
- Particle Size:−325 mesh