Order Entry
Germany
ContactUsLinkComponent
Potassium carbonate
Potassium carbonate
Catalog # OVER813100-25
CAS Number:  
undefined
Potassium carbonate
Catalog # OVER813100-25
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
EINECS:209-529-3
 :GHS07
Storage Temperature:Ambient
CAS Number:584-08-7
Formula:K₂CO₃
MDL Number:MFCD00011382