Order Entry
Germany
ContactUsLinkComponent
Sodium chloride, Sigma-Aldrich®
Sodium chloride, Sigma-Aldrich®
Catalog # SIALS6150-100TAB
Supplier:  Merck
CAS Number:  
Sodium chloride, Sigma-Aldrich®
Catalog # SIALS6150-100TAB
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0945
  • Pk Info:
    100 tablets

Specifications

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477