Order Entry
Germany
ContactUsLinkComponent
Sodium chloride
Sodium chloride
Catalog # DOWCSALTGRAN25
Supplier:  DOW CORNING
CAS Number:  
Sodium chloride
Catalog # DOWCSALTGRAN25
Supplier:  DOW CORNING
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477