Sodium bromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7647-15-6
:
Formula:NaBr
MDL Number:MFCD00003475
:102.89 g/mol
Merck Index:13,08666
Boiling Pt:1396 °C (1013 hPa)
Melting Pt:753 °C
Density:3.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-599-9