Order Entry
Germany
ContactUsLinkComponent
Sodium bromide
Sodium bromide
Catalog # PROONIST-977
CAS Number:  
Sodium bromide
Catalog # PROONIST-977
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :102.89 g/mol
Merck Index:13,08666
Boiling Pt:1396 °C (1013 hPa)
Melting Pt:753 °C
Density:3.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-599-9
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475