
di-Sodium tetraborate, anhydrous analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1330-43-4
:
Formula:Na₂B₄O₇
MDL Number:MFCD00081185
:201.22 g/mol
Merck Index:12,08733
Boiling Pt:1575 °C (1013 hPa)
Melting Pt:742 °C
Density:2.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08