1,4-Dioxane ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:100…102 °C (1.013 hPa)
Melting Pt:12 °C
Density:1.031…1.034 g/cm³ (20 °C)
Flash Pt:22.5 %(V)
Storage Temperature:Ambient
EINECS:204-661-8
UN:1165
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:complying
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Congealing - Freezing Point:Min. 11.0 °C
- Non-volatile matter:Max. 0.005 %
- Water (Karl Fischer):Max. 0.05 %
- Titrable Acid:Max. 0.0016 meq/g
- Carbonyl Compounds (as HCHO):Max. 0.01 %
- Peroxides (as H₂O₂):Max. 0.005 %
- APHA:Max. 20
- Explosion Limit:22 %
- Refractive Index:n20/D 1.422 (lit.)
- Autoignition Temperature:356 °F