Propylene carbonate for synthesis, Sigma-Aldrich®
: Merck
: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester
:108-32-7
:
:C₄H₆O₃
:MFCD00005385
:102.09 g/mol
:241…242 °C (1013 hPa)
:–48.8 °C
:1.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:132 °C (closed cup)
:203-572-1
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:240 g/l (20 °C)
- Melting Point:-49 °C
- Molar Mass:102.09 g/mol
- Boiling Point:243 °C (1013 hPa)
- Vapor Pressure:0.04 hPa (20 °C)
- Flash Point:123 °C
- Refractive Index:1.4209 (20 °C, 589 nm)
- Explosion Limit:1.8 - 14.3 %(V)
- Density:1.20 g/cm3 (20 °C)
- pH:7.0 (200 g/l, H2O, 20 °C)
- Ignition Point:435 °C DIN 51794
- Viscosity Kinematic:1.4 mm2/s (43 °C)