Propylene carbonate 99% (by GC)
About this item
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries.
It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Caution: Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
- CAS Number:108-32-7
- Formula:C₄H₆O₃
- MDL Number:MFCD00005385
- Molecular Weight:102.09 g/mol
- Boiling Pt:240…243 °C
- Melting Pt:–49 °C
- Density:1.204
- Flash Pt:132 °C (270 °F)
- Storage Temperature:Ambient
- EINECS:203-572-1
4 Options Available Below
Product Details & Documents
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries.
It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Caution: Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
- CAS Number:108-32-7
- Formula:C₄H₆O₃
- MDL Number:MFCD00005385
- Molecular Weight:102.09 g/mol
- Boiling Pt:240…243 °C
- Melting Pt:–49 °C
- Density:1.204
- Flash Pt:132 °C (270 °F)
- Storage Temperature:Ambient
- EINECS:203-572-1
Documents
Product Family Options
Product Information
- Pack typePk InfoPkAvailabilityPrice
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



