Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dibenzofuran
Dibenzofuran
Dibenzofuran
PROOBCR-337
:
LGC Standards PROMOCHEM
:
Dibenzofuran
PROOBCR-337
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
168.19 g/mol
:
285 °C (1013 hPa)
:
79…82 °C
:
0.966 g/cm³ (20 °C)
:
Refrigerator
:
205-071-3
:
132-64-9
:
C₁₂H₈O
:
MFCD00004968