Order Entry
Germany
ContactUsLinkComponent
2-Ethyl-9,10-anthraquinone for synthesis, Sigma-Aldrich®
Health
Environment
2-Ethyl-9,10-anthraquinone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Environment
2-Ethyl-9,10-anthraquinone for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.0007 g/l (20 °C)
  • Melting Point:
    107 - 110 °C
  • Molar Mass:
    236.26 g/mol
  • Bulk Density:
    650 kg/m3
  • Boiling Point:
    180 - 190 °C
  • Vapor Pressure:
    0.00056 hPa (70 °C)
  • Flash Point:
    210 °C
  • Ignition Point:
    485 °C
  • Hill Formula:
    C16H12O2

Specification Test Results

Related Information