9,10-Dihydroanthracene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:613-31-0
:
Formula:C₁₄H₁₂
MDL Number:MFCD00001239
:180.25 g/mol
Boiling Pt:312 °C (1013 hPa)
Melting Pt:70…72 °C
Density:1.207 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:210-336-1