Order Entry
Germany
ContactUsLinkComponent
 
1,8-Dihydroxy-9,10-anthraquinone for synthesis, Sigma-Aldrich®
  8.22028.9025
 :  Merck
 :  
1,8-Dihydroxy-9,10-anthraquinone for synthesis, Sigma-Aldrich®
  8.22028.9025
 :  Merck
 :  

 

  • Pk:
    25 kg
  • Seq:
    4283

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    190 - 193 °C
  • Molar Mass:
    240.21 g/mol
  • Bulk Density:
    600 kg/m3
  • Flash Point:
    200 °C

 

 

 :
 :240.22 g/mol
 :13,02841
 :190…193 °C
 :1.537 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :>200 °C
 :204-173-5
 :117-10-2
 :C₁₄H₈O₄
 :MFCD00001211