Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diamino-9,10-anthraquinone
1,4-Diamino-9,10-anthraquinone for synthesis, Sigma-Aldrich®
1,4-Diamino-9,10-anthraquinone for synthesis, Sigma-Aldrich®
:
Merck
1,4-Diamino-9,10-anthraquinone for synthesis, Sigma-Aldrich®
:
Merck
:
128-95-0
:
:
C₁₄H₁₀N₂O₂
:
MFCD00001224
:
238.25 g/mol
:
M-Clarity™ quality level = MQ200
:
204-922-6
:
GHS07
:
Ambient
msds_842128