9,10-Anthraquinone
Catalog # EHERC10281000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:208.22 g/mol
Merck Index:13,00692
Boiling Pt:380 °C (1013 hPa)
Melting Pt:165…169 °C
Density:1.419…1.438 g/cm³ (20 °C)
Flash Pt:185 °C
EINECS:201-549-0
CAS Number:84-65-1
Formula:C₁₄H₈O₂
MDL Number:MFCD00001188