Order Entry
Germany
ContactUsLinkComponent
Dibenzosuberone
Dibenzosuberone
Catalog # MOLE31304727-100G
Supplier:  Molekula
CAS Number:  
Dibenzosuberone
Catalog # MOLE31304727-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :208.26 g/mol
Boiling Pt:378 °C (1013 hPa)
Melting Pt:31…34 °C
Density:1.1635 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-912-3
CAS Number:1210-35-1
Formula:C₁₅H₁₂O
MDL Number:MFCD00003587