2,4,6,8-Tetramethylcyclotetrasiloxane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2370-88-9
:
Formula:C₄H₁₆O₄Si₄
MDL Number:MFCD00039567
Boiling Pt:142 °C (1013 hPa)
Melting Pt:–69 °C
Density:0.99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:240.51 g/mol
EINECS:219-137-4
Frequently Bought Together