2,2,4,4,6,6-Hexamethylcyclotrisilazane
Catalog # ABCRAB111169.0100
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:219.51 g/mol
Boiling Pt:186…188 °C (1013 hPa)
Density:0.9196 g/cm³ (20 °C)
EINECS:213-773-6
CAS Number:1009-93-4
Formula:C₆H₂₁N₃Si₃
MDL Number:MFCD00039698