3-Methyl-2-cyclopenten-1-one, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2758-18-1
:
Formula:C₆H₈O
MDL Number:MFCD00001403
:96.13 g/mol
Boiling Pt:158 °C (1013 hPa)
Melting Pt:2 °C
Density:0.97 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:220-421-5