Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cyclohexane, extra pure

Flammable
Health
Irritant
Environment
Supplier:   Bernd Kraft
Cyclohexane, extra pure
Supplier:  Bernd Kraft
Certificates
Cyclohexane, extra pure
Cyclohexane, extra pure

6 Options Available Below

Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,70 €
    32,70 €
    1 * 500 mL
       32,70 €

  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 70,30 €
    70,30 €
    1 * 2.500 mL
       70,30 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 38,80 €
    38,80 €
    1 * 1 L
       38,80 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 110,00 €
    110,00 €
    1 * 5 L
       110,00 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 180,00 €
    180,00 €
    1 * 10 L
       180,00 €

  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 394,00 €
    394,00 €
    1 * 25 L
       394,00 €