Cyclohexane
150 kg
Catalog # KRAE109930-9150
Supplier: KRAEMER MARTIN
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0.7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Specifications
Specifications
- Pk:150 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



