cis-1,2-Cyclohexanediol, Sigma-Aldrich®
Catalog # SIAL361267-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:116.16 g/mol
Boiling Pt:120 °C (15 mmHg)
Melting Pt:98…101 °C
Density:1.183 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
CAS Number:1792-81-0
Formula:C₆H₁₂O₂
MDL Number:MFCD00064944