Iodocyclohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:626-62-0
:
Formula:C₆H₁₁I
MDL Number:MFCD00003826
:210.06 g/mol
Boiling Pt:80 °C (1013 hPa)
Density:1.625 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:210-957-8