Bromocyclohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-85-0
:
Formula:C₆H₁₁Br
MDL Number:MFCD00003819
:163.06 g/mol
Merck Index:13,02759
Boiling Pt:166 °C (1013 hPa)
Melting Pt:–56.5 °C
Density:1.33 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:63 °C
Storage Temperature:Ambient
EINECS:203-622-2