Vitamin B12 (cyanocobalamin)
Catalog # MOLEM18601401
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1355.38 g/mol
Merck Index:12,10152
Melting Pt:320 °C
Storage Temperature:Refrigerator
EINECS:200-680-0
CAS Number:68-19-9
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092