Vitamin B12 (cyanocobalamin) 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68-19-9
:
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092
:1355.38 g/mol
Merck Index:12,10152
Melting Pt:320 °C
Storage Temperature:Ambient
EINECS:200-680-0
Specification Test Results
- Appearance:Red to dark red Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤12 % (20 mg, 105°C, 2 hrs) (vacuum)
- UV spectrophotometry:≥95 % (on dried substance)
- Ratio of Absorbances:3.15 to 3.45 (A361/A(547 to 559))