ContactUsLinkComponent
Chrysene
  PROOBCR-269
 :  
undefined
Chrysene
  PROOBCR-269
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    20 mg
  • Seq:
    2305
 :
 :228.29 g/mol
 :13,02275
 :448 °C (1013 hPa)
 :250…252 °C
 :1.274 g/cm³ (20 °C)
 :205-923-4
 :2811
 :6.1,III
 :GHS08
 :GHS09
 :218-01-9
 :C₁₈H₁₂
 :MFCD00003698