Order Entry
Germany
Orders LinkContactUsLinkComponent
Quinoxaline

Quinoxaline

Glass bottle for solids, 100 g
Catalog # MOLE28973649-100G
Marketsource
Supplier:   Molekula
Quinoxaline
Catalog # MOLE28973649-100G
Supplier:  Molekula
Marketsource
CAS Number:  91-19-0
Quinoxaline

Glass bottle for solids, 100 g

CAS Number:  91-19-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:91-19-0
  • Formula:C₈H₆N₂
  • MDL Number:MFCD00006719
  • Molecular Weight:130.15 g/mol
  • Merck Index:12,08263
  • Boiling Pt:230 °C (1013 hPa)
  • Melting Pt:30…32 °C
  • Density:1.1485 g/cm³ (32 °C)
  • EINECS:202-047-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.