



cis-Decahydronaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:493-01-6
:
Formula:C₁₀H₁₈
MDL Number:MFCD00064189
:138.25 g/mol
Merck Index:13,02866
Boiling Pt:194…196 °C (1013 hPa)
Melting Pt:–43 °C
Density:0.89 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-770-9
UN:1147
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:57 °C
:GHS02
:GHS05
:GHS07
:GHS09
Frequently Bought Together