Order Entry
Germany
ContactUsLinkComponent
cis-Decahydronaphthalene
Flammable
Corrosive
Irritant
Environment
cis-Decahydronaphthalene
Catalog # EHERC12094000
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Corrosive
Irritant
Environment
cis-Decahydronaphthalene
Catalog # EHERC12094000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :138.25 g/mol
Merck Index:13,02866
Boiling Pt:194…196 °C (1013 hPa)
Melting Pt:–43 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:207-770-9
UN:1147
ADR:3,III
 :GHS02
 :GHS05
 :GHS07
 :GHS09
CAS Number:493-01-6
Formula:C₁₀H₁₈
MDL Number:MFCD00064189