Bicyclo[3.3.1]-9-nonanone, Sigma-Aldrich®
Catalog # SIAL186015-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:138.21
Melting Pt:156…158 °C
:M-Clarity™ quality level = MQ100
EINECS:241-868-2
CAS Number:17931-55-4
Formula:C₉H₁₄O
MDL Number:MFCD00074752