Order Entry
Germany
ContactUsLinkComponent
5-Methylbenzotriazole
5-Methylbenzotriazole
Catalog # MOLE28528039-100G
Supplier:  Molekula
CAS Number:  
5-Methylbenzotriazole
Catalog # MOLE28528039-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:361 °C (1013 hPa)
Boiling Pt:361 °C (1013 hPa)
Melting Pt:85 °C
 :133.15 g/mol
EINECS:205-265-8
CAS Number:136-85-6
Formula:C₇H₇N₃
MDL Number:MFCD00005702