Order Entry
Germany
Orders LinkContactUsLinkComponent
Benzo[a]pyrene

Benzo[a]pyrene

100 mg, 1
Catalog # 48564.
Health
Environment
Supplier:   Merck
Benzo[a]pyrene
Catalog # 48564.
Supplier:  Merck
CAS Number:  50-32-8
Benzo[a]pyrene
Benzo[a]pyrene

100 mg, 1

CAS Number:  50-32-8
Price
240,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Synonyms:  Benzo[def]chrysene, BaP
  • CAS Number:50-32-8
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00003602
  • Molecular Weight:252.32 g/mol
  • Merck Index:12,01134
  • Boiling Pt:495 °C (1013 hPa)
  • Melting Pt:176…178 °C
  • Density:1.351 g/cm³ (20 °C)
  • EINECS:200-028-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    1
  • Pk Info:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.