Orders LinkContactUsLinkComponent
Benzo[a]pyrene ≥96% | Avantor
Benzo[a]pyrene ≥96%
Benzo[a]pyrene ≥96%
Benzo[a]pyrene ≥96%
Benzo[a]pyrene ≥96%
Purity: 96% CAS No.: 50-32-8 Molecular Formula: C20H12 Formula Weight: 252.30 Physical Form: Crystalline Appearance: Pale yellow/green/orange Melting Point: 177-180° Boiling Point: 495° Solubility: Soluble in benzene, toluene, and xylene. Sparingly soluble in alcohol, methanol. Insoluble in water Merck Reference: 14,1103 UN No.: 3077 MDL No.: MFCD00003602
Extensively used in cancer research
 :  Benzo[def]chrysene, BaP
  •  :50-32-8
  •  :C₂₀H₁₂
  •  :MFCD00003602
  •  :252.32 g/mol
  •  :14,1103
  •  :495 °C
  •  :177…180 °C
  •  :1.351 g/cm³ (20 °C)
  •  :Ambient
  •  :200-028-5
  •  :3077
  •  :9,III
Dokumente
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    100 mg
    1 * 100 mg
    1 * 100 mg - 68,70 €
    68,70 €
    1 * 100 mg
       68,70 €
  • Glass bottle for solids
    0,5 g
    1 * 0,5 g
    1 * 0,5 g - 277,00 €
    277,00 €
    1 * 0,5 g
       277,00 €