7-Amino-4-methylcoumarin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:26093-31-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00006868
:175.19 g/mol
Boiling Pt:352 °C (1013 hPa)
Melting Pt:226 °C
Density:1.377 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:247-454-8