ContactUsLinkComponent
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin 98%
Certificates
undefined
7-Amino-4-methylcoumarin 98%
Specification Test Results
  • Appearance:
    Yellow to beige to brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %