Order Entry
Germany
ContactUsLinkComponent
 
4-Hydroxycoumarin
Catalog # MOLE36136514-500G
Supplier:  Molekula
CAS Number:  
4-Hydroxycoumarin
Catalog # MOLE36136514-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Boiling Pt:349 °C (1013 hPa)
Melting Pt:211…214 °C
 :162.14 g/mol
EINECS:214-060-2
CAS Number:1076-38-6
Formula:C₉H₆O₃
MDL Number:MFCD00006856