

Biphenyl (PCB No. 0), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:92-52-4
:
Formula:C₁₂H₁₀
MDL Number:MFCD00003054
:154.21 g/mol
Merck Index:13,03346
Boiling Pt:255 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.175 g/cm³ (20 °C)
EINECS:202-163-5
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
Flash Pt:113 °C
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together