

Biphenyl (PCB No. 0)
Artikelnr. PROOLGC2610
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:154.21 g/mol
Merck Index:13,03346
Boiling Pt:255 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.175 g/cm³ (20 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:202-163-5
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:92-52-4
Formula:C₁₂H₁₀
MDL Number:MFCD00003054