

Biphenyl (PCB No. 0) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-52-4
:
Formula:C₁₂H₁₀
MDL Number:MFCD00003054
:154.21 g/mol
Merck Index:13,03346
Boiling Pt:255 °C (760 mmHg)
Melting Pt:68…72 °C
Density:1,175 g/cm³ (20 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:202-163-5
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:68 to 72 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together