2,2',3,4',5,6-Hexachlorobiphenyl (PCB No. 147)
Catalog # PROOCIL-PCB-147-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
CAS Number:68194-13-8
Formula:C₁₂H₄Cl₆