2,2',5,5'-Tetrachlorobiphenyl (PCB No. 52)
Catalog # EHERC20005200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:291.99 g/mol
Storage Temperature:Refrigerator
UN:2315
ADR:9,II
CAS Number:35693-99-3
Formula:C₁₂H₆Cl₄